Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-cyanopyridine, 97%, Thermo Scientific™

Catalog No. AAH32550MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32550MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32550MD Supplier Thermo Scientific Chemicals Supplier No. H32550MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78790-79-1
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD04035596
InChI Key PEZNQSQPDQLHPN-UHFFFAOYSA-N
Synonym 3-amino-4-cyanopyridine, 3-aminoisonicotinonitrile, 3-amino-isonicotinonitrile, 4-pyridinecarbonitrile, 3-amino, pubchem9759, 3-amino4-cyanopyridine, acmc-209pfc, 3-amino-4-cyano-pyridine, 3-aminopyridine4-carbonitrile, ksc497q6t
PubChem CID 10678219
IUPAC Name 3-aminopyridine-4-carbonitrile
SMILES NC1=C(C=CN=C1)C#N

Specifications

Melting Point 135°C to 138°C
Quantity 250 mg
UN Number UN3439
Formula Weight 119.13
Percent Purity 97%
Chemical Name or Material 3-Amino-4-cyanopyridine
Hazard Category H301+H311-H319
Hazard Statement GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.