Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH62228MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62228MD 250 mg
AAH6222803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62228MD Supplier Thermo Scientific Chemicals Supplier No. H62228MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003575-43-6
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD11044430
InChI Key VXNIVUKZNDRJBW-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-4-fluorophenylboronic acid, pinacol ester, 3-amino-4-fluorophenylboronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 3-amino-4-fluorobenzeneboronic acid pinacol ester, 3-amino-4-fluorophenyl boronic acid pinacol ester, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
PubChem CID 46737990
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC(N)=C(F)C=C1

Specifications

Quantity 250 mg
Formula Weight 237.08
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Amino-4-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.