Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-fluorophenylboronic acid, 97%

Catalog No. AC445290010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445290010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445290010 Supplier Thermo Scientific Chemicals Supplier No. 445290010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 873566-75-7 | C6H7BFNO2 | 154.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873566-75-7
Molecular Formula C6H7BFNO2
Molecular Weight (g/mol) 154.94
MDL Number MFCD03092925
InChI Key SYBMNJPUZMUPGQ-UHFFFAOYSA-N
Synonym 3-amino-4-fluorophenyl boronic acid, 3-amino-4-fluorobenzeneboronic acid, 5-borono-2-fluoroaniline, 3-amino-4-fluoro-phenyl boronic acid, boronic acid, 3-amino-4-fluorophenyl, boronic acid,b-3-amino-4-fluorophenyl, hydrochloride, pubchem1743, acmc-209qjr, 3-amino-4-fluoro phenylboronic acid
PubChem CID 21954814
IUPAC Name (3-amino-4-fluorophenyl)boronic acid
SMILES NC1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 226°C to 232°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 154.94
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Amino-4-fluorophenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.