Learn More
Apexbio Technology LLC 3-AMINO-4-HYDROXYBENZOIC-ACID
SDP
Catalog No. 5000568368
Encompass_Preferred
3-Amino-4-hydroxybenzoic acid (CAS No 1571-72-8) is a compound of interest in biomedical research due to its structural attributes and potential biological activities Originating from the benzoic acid family it plays a role in pathways involving aromatic amino acids The compound often targets specific cellular pathways influencing enzyme activities and cell signaling mechanisms In vitro it demonstrates activity typically in the nanomolar to low micromolar range making it suitable for early-stage screening assays Its applications extend to various research areas including cell model studies and preliminary animal experiments It serves as a probe in drug screening to evaluate pharmacodynamic responses Experimental concentrations typically depend on the specific research design but they must be carefully calibrated according to the desired outcome and model system Understanding its mechanism is critical for applications in pharmaceuticals and therapeutic development
Catalog No. 50-005-68368 Supplier Apexbio Technology LLC Supplier No. C6908100
3-Amino-4-hydroxybenzoic acid (CAS No 1571-72-8) is a compound of interest in biomedical research due to its structural attributes and potential biological activities Originating from the benzoic acid family it plays a role in pathways involving aromatic amino acids The compound often targets specific cellular pathways influencing enzyme activities and cell signaling mechanisms In vitro it demonstrates activity typically in the nanomolar to low micromolar range making it suitable for early-stage screening assays Its applications extend to various research areas including cell model studies and preliminary animal experiments It serves as a probe in drug screening to evaluate pharmacodynamic responses Experimental concentrations typically depend on the specific research design but they must be carefully calibrated according to the desired outcome and model system Understanding its mechanism is critical for applications in pharmaceuticals and therapeutic development