Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-4-hydroxybenzoic acid | 1571-72-8 | MFCD00007697 | 99.9% | 153.14 | 1 G
SDP

Catalog No. 5000453994
Encompass_Preferred

3-Amino-4-hydroxybenzoic acid is an endogenous metabolite and a monohydroxybenzoic acid. It features an amino substituent at position 3 of 4-hydroxybenzoic acid and is classified as an aminobenzoic acid and a conjugate acid.

  • Endogenous metabolite
  • Monohydroxybenzoic acid
  • Aminobenzoic acid
  • Conjugate acid
  • Off-white to light brown solid
  • Purity 99.86%

Catalog No. 50-004-53994 Supplier Medchemexpress LLC Supplier No. HYW0102241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-4-hydroxybenzoic acid is an endogenous metabolite and a monohydroxybenzoic acid. It features an amino substituent at position 3 of 4-hydroxybenzoic acid and is classified as an aminobenzoic acid and a conjugate acid.

  • Endogenous metabolite
  • Monohydroxybenzoic acid
  • Aminobenzoic acid
  • Conjugate acid
  • Off-white to light brown solid
  • Purity 99.86%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.