Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methoxybenzenesulfonic acid, 98%, Thermo Scientific™

Catalog No. AC342610250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC342610250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC342610250 Supplier Thermo Scientific Chemicals Supplier No. 342610250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-42-0
Molecular Formula C7H9NO4S
Molecular Weight (g/mol) 203.212
MDL Number MFCD00035759
InChI Key FLIOATBXVNLPLK-UHFFFAOYSA-N
Synonym 4-methoxymetanilic acid, 2-anisidine-4-sulfonic acid, 2-methoxyaniline-5-sulfonic acid, benzenesulfonic acid, 3-amino-4-methoxy, o-anisidine-4-sulfonic acid, 2-methoxy-5-sulfoaniline, o-anisidine-p-sulfonic acid, 2-aminoanisole-4-sulfonic acid, benzenesulfonic acid, 3-amino-4-methoxyl, acmc-20ahhh
PubChem CID 66821
IUPAC Name 3-amino-4-methoxybenzenesulfonic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H3COC6H3(NH2)SO2OH
Quantity 25 g
Formula Weight 203.21
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybenzenesulfonic acid, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-667-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.