Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-4-pyrazolecarbonitrile | 16617-46-2 | 108.10 | 500 G
SDP

Catalog No. 5000447087
Encompass_Preferred

3-Amino-4-pyrazolecarbonitrile is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical reagent
  • Biological material
  • Organic compound for life science related research
  • Purity of 98.0%
  • Molecular weight of 108.10
  • For research use only

Catalog No. 50-004-47087 Supplier Medchemexpress LLC Supplier No. HYW001964500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-4-pyrazolecarbonitrile is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical reagent
  • Biological material
  • Organic compound for life science related research
  • Purity of 98.0%
  • Molecular weight of 108.10
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.