Learn More
Medchemexpress LLC 3-Amino-5-bromo-2-methylpyridine | 914358-73-9 | 187.04 | 25 G
SDP
Catalog No. 5000449477
Encompass_Preferred
3-Amino-5-bromo-2-methylpyridine (3ABMP) is a biochemical reagent. It should be stored at 4°C, protected from light. In solvent, it should be stored at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Biochemical reagent
  • Suitable for biochemical assays
  • Storage at 4°C, protected from light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-49477 Supplier Medchemexpress LLC Supplier No. 25GHYW003575
Add to Cart
Add to Cart
3-Amino-5-bromo-2-methylpyridine (3ABMP) is a biochemical reagent. It should be stored at 4°C, protected from light. In solvent, it should be stored at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Biochemical reagent
  • Suitable for biochemical assays
  • Storage at 4°C, protected from light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light