Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-5-bromopyridine | 13535-01-8 | 99.68% | C5H5BrN2 | 10 G
SDP

Catalog No. 5000434232
Encompass_Preferred

3-Amino-5-bromopyridine is a solid chemical compound with the molecular formula C5H5BrN2. It is described as an intermediate and antibacterial agent, characterized by its high purity and specific physical properties.

  • Appearance: Solid
  • Purity: 99.68%
  • CAS number: 13535-01-8
  • Molecular formula: C5H5BrN2
  • Molecular weight: 173.01
  • Melting/freezing point: 65-69°C
  • Boiling point: 149-150 °C at 12 mmHg

Catalog No. 50-004-34232 Supplier Medchemexpress LLC Supplier No. HY6605210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-5-bromopyridine is a solid chemical compound with the molecular formula C5H5BrN2. It is described as an intermediate and antibacterial agent, characterized by its high purity and specific physical properties.

  • Appearance: Solid
  • Purity: 99.68%
  • CAS number: 13535-01-8
  • Molecular formula: C5H5BrN2
  • Molecular weight: 173.01
  • Melting/freezing point: 65-69°C
  • Boiling point: 149-150 °C at 12 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.