Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-5-bromopyridine | 13535-01-8 | 99.7% | C5H5BrN2 | 25 G
SDP

Catalog No. 5000358969
Encompass_Preferred

MedChemExpress HY-66052 is 3-Amino-5-bromopyridine, a high-purity chemical compound (99.7% GC) identified by CAS number 13535-01-8. This versatile compound serves as an intermediate and antibacterial agent, crucial for various applications including organic synthesis, pharmaceutical development, agrochemical production, and the manufacturing of electronic components and perovskite solar cells. It appears as a light yellow to brown solid and should be stored at 4°C, protected from light.

  • Intermediate and antibacterial agent
  • Utilized in organic synthesis
  • Applied in pharmaceuticals and agrochemicals
  • Essential for electronic components
  • Used in perovskite solar cells
  • Purity of 99.7% (GC)
  • Light yellow to brown solid appearance
  • Store at 4°C, protect from light

Catalog No. 50-003-58969 Supplier Medchemexpress LLC Supplier No. 25GHY66052
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MedChemExpress HY-66052 is 3-Amino-5-bromopyridine, a high-purity chemical compound (99.7% GC) identified by CAS number 13535-01-8. This versatile compound serves as an intermediate and antibacterial agent, crucial for various applications including organic synthesis, pharmaceutical development, agrochemical production, and the manufacturing of electronic components and perovskite solar cells. It appears as a light yellow to brown solid and should be stored at 4°C, protected from light.

  • Intermediate and antibacterial agent
  • Utilized in organic synthesis
  • Applied in pharmaceuticals and agrochemicals
  • Essential for electronic components
  • Used in perovskite solar cells
  • Purity of 99.7% (GC)
  • Light yellow to brown solid appearance
  • Store at 4°C, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.