Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-chloropyridine, 95%

Catalog No. AC448900050
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448900050 5 g Glass bottle
AC448900010 1 g Glass bottle
AC448902500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC448900050 Supplier Thermo Scientific Chemicals Supplier No. 448900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 22353-34-0 | C5H5ClN2 | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22353-34-0
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
InChI Key ZHMASVAJFJFFLS-UHFFFAOYSA-N
PubChem CID 818257
IUPAC Name 5-chloropyridin-3-amine
SMILES C1=C(C=NC=C1Cl)N

Specifications

Melting Point 51.0°C to 56.0°C
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 3-amino-5-chloropyridine, 5-chloro-3-pyridinamine, 5-chloro-pyridin-3-ylamine, 3-pyridinamine, 5-chloro, 5-chloro-3-pyridylamine, 5-chloro-3-aminopyridine, 5-chloropyridine-3-amine, 3-pyridinamine,5-chloro, 5-chloro-3-pyridinylamine, 3-amino-5-chloro-pyridine
Solubility Information Solubility in water: insoluble
Formula Weight 128.56
Percent Purity 95%
Chemical Name or Material 3-Amino-5-chloropyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.