Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-ethyl-1H-pyrazole, 97%

Catalog No. AC446710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446710010 1 g
AC446710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446710010 Supplier Thermo Scientific Chemicals Supplier No. 446710010
Add to Cart

CAS: 1904-24-1 | C5H9N3 | 111.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1904-24-1
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD08060982
InChI Key AXDGPQLEVYSXNL-UHFFFAOYSA-N
Synonym 3-ethyl-1h-pyrazol-5-amine, 5-amino-3-ethyl-1h-pyrazole, 3-amino-5-ethylpyrazole, 3-amino-5-ethyl-1h-pyrazole, 5-ethyl-2h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 5-ethyl, 5-ethyl-2h-pyrazol-3-amine, ethyl-5-aminopyrazole, 3-amino-5-ethypyrazole, 1h-pyrazol-5-amine, 3-ethyl
PubChem CID 21991095
IUPAC Name 5-ethyl-1H-pyrazol-3-amine
SMILES CCC1=CC(N)=NN1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material 3-Amino-5-ethyl-1H-pyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.