Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-hydroxy-1H-pyrazole, 98%

Catalog No. p-7081433
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1141206 5 g
AAA1141214 25 g
AAA1141222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1141206 Supplier Thermo Scientific Chemicals Supplier No. A1141206
Only null left
Add to Cart

CAS: 6126-22-3 | C3H5N3O | 99.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6126-22-3
Molecular Formula C3H5N3O
Molecular Weight (g/mol) 99.093
MDL Number MFCD00022384
InChI Key QZBGOTVBHYKUDS-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxypyrazole, 3-amino-1h-pyrazol-5-ol, 5-amino-1h-pyrazol-3-ol, 5-amino-1h-pyrazol-3 2h-one, 3-amino-1,2-dihydropyrazol-5-one, 5-aminopyrazol-3-ol, 5-amino-2h-pyrazol-3-ol, 3-pyrazolin-5-one, 3-amino, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 1h-pyrazol-5-ol, 3-amino
PubChem CID 96221
IUPAC Name 5-amino-1,2-dihydropyrazol-3-one
SMILES C1=C(NNC1=O)N

Specifications

Melting Point ∼210°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 507793
Formula Weight 99.09
Percent Purity 98%
Chemical Name or Material 3-Amino-5-hydroxy-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-095-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.