Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-amino-5-hydroxybenzoic acid | 76045-71-1 | MFCD06797571 | 99.8% | 153.14 g/mol | C7H7NO3 | 250 MG
SDP

Catalog No. 5000208691
Encompass_Preferred

3-Amino-5-hydroxybenzoic acid is a research-grade small molecule used as a biosynthetic precursor and drug intermediate in studies of ansamycin antibiotics. It is supplied at high purity and is available in multiple pack sizes for laboratory use.

  • Used as a biosynthetic precursor in ansamycin antibiotic research.
  • Purity 99.8%.
  • Molecular formula C7H7NO3.
  • Molecular weight 153.14 g/mol.
  • CAS number 76045-71-1.
  • Available in multiple pack sizes, including 250 MG.

Catalog No. 50-002-08691 Supplier Medchemexpress LLC Supplier No. 250MGHYW077634
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-5-hydroxybenzoic acid is a research-grade small molecule used as a biosynthetic precursor and drug intermediate in studies of ansamycin antibiotics. It is supplied at high purity and is available in multiple pack sizes for laboratory use.

  • Used as a biosynthetic precursor in ansamycin antibiotic research.
  • Purity 99.8%.
  • Molecular formula C7H7NO3.
  • Molecular weight 153.14 g/mol.
  • CAS number 76045-71-1.
  • Available in multiple pack sizes, including 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.