Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-methylisoxazole 97.0+%, TCI America™
SDP

Catalog No. A098025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A098025G 25 g
A0980250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A098025G Supplier TCI America Supplier No. A098025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1072-67-9
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.11
MDL Number MFCD00003155
InChI Key FKPXGNGUVSHWQQ-UHFFFAOYSA-N
Synonym 3-amino-5-methylisoxazole, 5-methylisoxazol-3-amine, 5-methyl-3-isoxazolamine, 3-isoxazolamine, 5-methyl, 5-methyl-3-aminoisoxazole, isoxazole, 3-amino-5-methyl, 5-methylisoxazol-3-ylamine, 3-amino-5-methyl-isoxazole, unii-g54mjs11l9, 3-amino-5-methylisoxazol
PubChem CID 66172
IUPAC Name 5-methyl-1,2-oxazol-3-amine
SMILES CC1=CC(N)=NO1

Specifications

Melting Point 63°C
Quantity 25 g
Formula Weight 98.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-methylisoxazole
EINECSNumber (9)-2177
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.