Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Amino-5-methylpyrazole | 31230-17-8 | 98.7% | C4H7N3 | 100 G
SDP

Catalog No. 5000433917
Encompass_Preferred

3-Amino-5-methylpyrazole is a pyrazole heterocyclic compound that serves as a key intermediate for the synthesis of pyrazolylamide derivatives. It appears as a solid below 41°C and a liquid above 43°C, with a colorless to light yellow appearance. This chemical has a molecular weight of 97.12.

  • Serves as a key intermediate for pyrazolylamide derivatives synthesis
  • Appears as solid below 41°C and liquid above 43°C
  • Has a colorless to light yellow appearance
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-33917 Supplier Medchemexpress LLC Supplier No. HY75637100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-5-methylpyrazole is a pyrazole heterocyclic compound that serves as a key intermediate for the synthesis of pyrazolylamide derivatives. It appears as a solid below 41°C and a liquid above 43°C, with a colorless to light yellow appearance. This chemical has a molecular weight of 97.12.

  • Serves as a key intermediate for pyrazolylamide derivatives synthesis
  • Appears as solid below 41°C and liquid above 43°C
  • Has a colorless to light yellow appearance
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.