Learn More
Medchemexpress LLC 3-amino-5-morpholinomethyl-2-oxazolidinone D5 | 1017793-94-0 | MFCD04973439 | 99.0% | C8H10D5N3O3 | 10MG
SDP
Catalog No. 5000261980
Encompass_Preferred
AMOZ-d5 is a deuterium-labeled analogue of AMOZ, the tissue-bound metabolite of the nitrofuran antibiotic furaltadone. It is provided as an isotopically labeled internal standard and tracer for accurate quantitative analysis in research and residue testing by GC-MS, LC-MS, and NMR.
  • Isotopically labeled internal standard for quantitative mass spectrometry and NMR.
  • Contains five deuterium atoms for a distinct mass shift from the native analyte.
  • Suitable for residue analysis and method validation in biological matrices.
  • High purity for accurate quantitation and reproducible results.
  • Stable under recommended storage conditions for reliable long-term use.
Catalog No. 50-002-61980 Supplier Medchemexpress LLC Supplier No. HY131144S10MG
AMOZ-d5 is a deuterium-labeled analogue of AMOZ, the tissue-bound metabolite of the nitrofuran antibiotic furaltadone. It is provided as an isotopically labeled internal standard and tracer for accurate quantitative analysis in research and residue testing by GC-MS, LC-MS, and NMR.
  • Isotopically labeled internal standard for quantitative mass spectrometry and NMR.
  • Contains five deuterium atoms for a distinct mass shift from the native analyte.
  • Suitable for residue analysis and method validation in biological matrices.
  • High purity for accurate quantitation and reproducible results.
  • Stable under recommended storage conditions for reliable long-term use.