Learn More
Medchemexpress LLC 3-Amino-5-morpholinomethyl-2-oxazolidone | 43056-63-9 | 99.9% | 250 MG
SDP
Catalog No. 5000341462
Encompass_Preferred
AMOZ is a carcinogenic metabolite of Furaltadone that forms stable tissue residues by covalently binding to proteins as a hapten. It can be coupled with carrier proteins to induce an immune response and is released after acid hydrolysis. AMOZ residues can be detected by competitive ELISA and can be used as a typical marker to monitor drug residues in animal-derived foods for food safety.
  • Forms stable tissue residues by covalently binding to proteins
  • Can be coupled with carrier proteins to induce an immune response
  • Released after acid hydrolysis
  • Residues detectable by competitive ELISA
  • Can be used as a marker to monitor drug residues in animal-derived foods
  • Shows high specificity in monoclonal antibody detection with a limit of detection as low as 0.16 μg/kg
  • Achieves enhanced sensitivity via chemiluminescence, detecting concentrations from 0.03-64 μg/l with a 0.01 μg/l limit of detection
Catalog No. 50-003-41462 Supplier Medchemexpress LLC Supplier No. 250MGHY131146
AMOZ is a carcinogenic metabolite of Furaltadone that forms stable tissue residues by covalently binding to proteins as a hapten. It can be coupled with carrier proteins to induce an immune response and is released after acid hydrolysis. AMOZ residues can be detected by competitive ELISA and can be used as a typical marker to monitor drug residues in animal-derived foods for food safety.
  • Forms stable tissue residues by covalently binding to proteins
  • Can be coupled with carrier proteins to induce an immune response
  • Released after acid hydrolysis
  • Residues detectable by competitive ELISA
  • Can be used as a marker to monitor drug residues in animal-derived foods
  • Shows high specificity in monoclonal antibody detection with a limit of detection as low as 0.16 μg/kg
  • Achieves enhanced sensitivity via chemiluminescence, detecting concentrations from 0.03-64 μg/l with a 0.01 μg/l limit of detection