Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-tert-butylpyrazole 98.0+%, TCI America™
SDP

Catalog No. A21011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21011G Supplier TCI America Supplier No. A21011G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82560-12-1
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD00067987,MFCD00082631
InChI Key ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
Synonym 3-amino-5-tert-butylpyrazole, 3-tert-butyl-1h-pyrazol-5-amine, 5-amino-3-tert-butylpyrazole, 3-amino-5-tert-butyl-1h-pyrazole, 5-tert-butyl-2h-pyrazol-3-ylamine, 5-amino-3-t-butylpyrazole, 1h-pyrazol-3-amine, 5-1,1-dimethylethyl, 5-tert-butyl-2h-pyrazol-3-amine, 5-amino-3-tert-butyl-1h-pyrazole
PubChem CID 522787
IUPAC Name 5-tert-butyl-1H-pyrazol-3-amine
SMILES CC(C)(C)C1=CC(N)=NN1

Specifications

Melting Point 78°C
Quantity 1 g
Formula Weight 139.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-tert-butylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.