Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-bromo-2-methylpyridine 97.0+%, TCI America™
SDP

Catalog No. A29285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29285G 5 g
A29281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29285G Supplier TCI America Supplier No. A29285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126325-47-1
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03095086
InChI Key UBTQJSZELCWGCN-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
PubChem CID 14418045
IUPAC Name 6-bromo-2-methylpyridin-3-amine
SMILES CC1=C(N)C=CC(Br)=N1

Specifications

Melting Point 110°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-6-bromo-2-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.