Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-amino-6-bromopyridine | 13534-97-9 | MFCD01646116 | 99.1% | 173.01 g/mol | C5H5BrN2 | 25 G
SDP

Catalog No. 5000302678
Encompass_Preferred

6-Bromopyridin-3-amine is an organic building block and research reagent used in medicinal chemistry and synthetic organic chemistry. Supplied as a high-purity solid, it serves as an intermediate for the preparation of brominated heterocycles and related derivatives, with documentation available for batch quality and safety.

  • High reported purity (~99.1%).
  • Molecular weight 173.01 g/mol.
  • Chemical formula C5H5BrN2.
  • Available in multiple lab-scale pack sizes for flexibility.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-003-02678 Supplier Medchemexpress LLC Supplier No. 25GHYW016849
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromopyridin-3-amine is an organic building block and research reagent used in medicinal chemistry and synthetic organic chemistry. Supplied as a high-purity solid, it serves as an intermediate for the preparation of brominated heterocycles and related derivatives, with documentation available for batch quality and safety.

  • High reported purity (~99.1%).
  • Molecular weight 173.01 g/mol.
  • Chemical formula C5H5BrN2.
  • Available in multiple lab-scale pack sizes for flexibility.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.