Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-amino-6-bromopyridine | 13534-97-9 | MFCD01646116 | 99.1% | 173.01 g/mol | C5H5BrN2 | 250 G
SDP

Catalog No. 5000303614
Encompass_Preferred

6-Bromopyridin-3-amine (CAS 13534-97-9) is an organic biochemical reagent used as a building block in medicinal chemistry and organic synthesis. It is supplied as a light brown to yellow solid with high chemical purity, and is suitable for use in synthetic intermediates, assay development, and other research applications.

  • High chemical purity (~99.1%) suitable for research and analytical use
  • Molecular formula C5H5BrN2 and molecular weight 173.01 g/mol for accurate stoichiometry
  • Solid, light brown to yellow form that is easy to handle and weigh
  • Available in multiple pack sizes including 250 g for scale-up needs
  • Store protected from light; in solvent, -80 °C (up to 6 months) or -20 °C (up to 1 month)

Catalog No. 50-003-03614 Supplier Medchemexpress LLC Supplier No. 250GHYW016849
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromopyridin-3-amine (CAS 13534-97-9) is an organic biochemical reagent used as a building block in medicinal chemistry and organic synthesis. It is supplied as a light brown to yellow solid with high chemical purity, and is suitable for use in synthetic intermediates, assay development, and other research applications.

  • High chemical purity (~99.1%) suitable for research and analytical use
  • Molecular formula C5H5BrN2 and molecular weight 173.01 g/mol for accurate stoichiometry
  • Solid, light brown to yellow form that is easy to handle and weigh
  • Available in multiple pack sizes including 250 g for scale-up needs
  • Store protected from light; in solvent, -80 °C (up to 6 months) or -20 °C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.