Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-L-tyrosine Dihydrochloride Monohydrate, Thermo Scientific™

Catalog No. AC159200250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159200250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159200250 Supplier Thermo Scientific Chemicals Supplier No. 159200250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23279-22-3
Molecular Formula C9H14Cl2N2O3
Molecular Weight (g/mol) 269.122
MDL Number MFCD00150105
InChI Key PIFWBMTWCTUOOQ-KLXURFKVSA-N
Synonym 3-amino-l-tyrosine dihydrochloride, h-tyr 3-nh2-oh.2hcl, s-2-amino-3-3-amino-4-hydroxyphenyl propanoic acid dihydrochloride, 2s-2-amino-3-3-amino-4-hydroxyphenyl propanoic acid dihydrochloride, h-3-amino-tyr-oh 2 hcl, m-nh2-tyr-oh.dihydrochloride, s-2-amino-3-3-amino-4-hydroxyphenyl-propanoic acid dihydrochloride
PubChem CID 46735196
IUPAC Name (2S)-2-amino-3-(3-amino-4-hydroxyphenyl)propanoic acid;dihydrochloride
SMILES C1=CC(=C(C=C1CC(C(=O)O)N)N)O.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC6H3(OH)CH2CH(NH2)CO2H·2HCl·H2O
Quantity 25 g
Beilstein 14, 622
Formula Weight 285.13
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-L-tyrosine dihydrochloride monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.