Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Amino-N,N-diethyl-propionamide hydrochloride

Catalog No. CDS005055 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005055 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005055 Supplier Sigma Aldrich Supplier No. CDS005055100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethly vinyl sulfone alkylates α-amino groups of lysine side chains and imidazole groups of histidine residues in proteins
  • Chemical modification of bovine serum albumin by ethyl vinyl sulfone has been studied by x-ray photoelectron spectroscopy

Specifications

CAS 34105-56-1
Molecular Formula C7H17ClN2O
Linear Formula C7H17ClN2O
MDL Number MFCD07366747
Quantity 5 g
Molecular Weight (g/mol) 180.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.