Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzaldehyde Polymer, TCI America™
SDP

Catalog No. A025925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A025925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A025925G Supplier TCI America Supplier No. A025925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29159-23-7
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00147530
InChI Key SIXYIEWSUKAOEN-UHFFFAOYSA-N
Synonym Poly(3-aminobenzaldehyde)
PubChem CID 74366
IUPAC Name 3-aminobenzaldehyde
SMILES C1=CC(=CC(=C1)N)C=O

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzaldehyde Polymer
EINECSNumber (3)-1163
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.