Learn More
Selleck Chemical LLC 3-Aminobenzamide S1132-25mg
SDP
Catalog No. 503108357
Encompass_Preferred
3-Aminobenzamide (3-ABA 3-Amino Benzamide 3-AB) is a potent inhibitor of Poly(ADP-ribose)polymerase (PARP) and inhibits cell apoptosis after SCI (Spinal Cord Injury) in caspase-independent way
Catalog No. 50-310-8357 Supplier Selleck Chemical LLC Supplier No. S113225MG
3-Aminobenzamide (3-ABA 3-Amino Benzamide 3-AB) is a potent inhibitor of Poly(ADP-ribose)polymerase (PARP) and inhibits cell apoptosis after SCI (Spinal Cord Injury) in caspase-independent way