Learn More
Enzo Life Sciences 3-Aminobenzamide (1g). CAS: 3544-24-9
SDP
Supplier:  Enzo Life Sciences ALX270044G001
Encompass_Preferred
Inhibits endogenous poly-ADP-ribosyltransferases often present in membrane preparations with minimal effect of bacterial toxin mediated ADP-ribosylation. Inhibits nitric oxide-induced apoptosis but not differentiation in HL-60 cells. Protects cells from oxygen radical and nitric oxide toxicity. Inhibits stress-induced apoptosis. Alternative name: 3-AB. Formula: C7H8N2O. MW: 136.2. Purity: ≥97% (GC). Appearance: White to tan powder. Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: Ambient.
Catalog No. 50-200-8378