Learn More
Medchemexpress LLC HY-12022 200mg Medchemexpress, 3-Aminobenzamide CAS:3544-24-9 Purity:>98%
SDP
Catalog No. 502080936
Encompass_Preferred
Medchemexpress, HY-12022 200mg 3-Aminobenzamide CAS:3544-24-9 3-Aminobenzamide (PARP-IN-1) is a potent inhibitor of PARP with IC50 of appr 50 nM in CHO cells, and acts as a mediator of oxidant-induced myocyte dysfunction during reperfusion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0936 Supplier Medchemexpress LLC Supplier No. HY12022200MG
Medchemexpress, HY-12022 200mg 3-Aminobenzamide CAS:3544-24-9 3-Aminobenzamide (PARP-IN-1) is a potent inhibitor of PARP with IC50 of appr 50 nM in CHO cells, and acts as a mediator of oxidant-induced myocyte dysfunction during reperfusion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.