Learn More
Apexbio Technology LLC 3-aminobenzamide 3544-24-9 500mg
SDP
Catalog No. 5000095587
Encompass_Preferred
3-Aminobenzamide (CAS 3544-24-9) is a potent inhibitor of poly(ADP-ribose) polymerase (PARP) exhibiting an IC50 of approximately 50 nM in CHO cell assays At concentrations above 1 M it suppresses over 95% of PARP activity without notable cytotoxicity In cellular models 3-aminobenzamide mitigates oxidative stress-induced endothelial dysfunction by enhancing acetylcholine-induced nitric oxide-mediated vasodilation following hydrogen peroxide exposure In vivo it has been shown to attenuate diabetic nephropathy in Leprdb/db mice by reducing albuminuria glomerular mesangial expansion and podocyte loss 3-Aminobenzamide serves as a valuable tool for PARP-related research in oxidative injury and metabolic disease models
Catalog No. 50-000-95587 Supplier Apexbio Technology LLC Supplier No. A4161500
3-Aminobenzamide (CAS 3544-24-9) is a potent inhibitor of poly(ADP-ribose) polymerase (PARP) exhibiting an IC50 of approximately 50 nM in CHO cell assays At concentrations above 1 M it suppresses over 95% of PARP activity without notable cytotoxicity In cellular models 3-aminobenzamide mitigates oxidative stress-induced endothelial dysfunction by enhancing acetylcholine-induced nitric oxide-mediated vasodilation following hydrogen peroxide exposure In vivo it has been shown to attenuate diabetic nephropathy in Leprdb/db mice by reducing albuminuria glomerular mesangial expansion and podocyte loss 3-Aminobenzamide serves as a valuable tool for PARP-related research in oxidative injury and metabolic disease models