Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminobenzamide | 3544-24-9 | >99.0% | 100 MG
SDP

Catalog No. 5000234214
Encompass_Preferred

3-Aminobenzamide is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) used in biochemical and cellular research to study DNA repair and oxidant-induced cellular responses. It is supplied as an analytical standard suitable for HPLC, GC, and MS applications.

  • Cas 3544-24-9.
  • Chemical formula C7H8N2O, molecular weight 136.15 g/mol.
  • Used as a PARP inhibitor in biochemical and cellular assays.
  • Supplied as an analytical standard for qualitative and quantitative analysis.
  • Available in multiple pack sizes including 100 MG.
  • Intended for research use only.

Catalog No. 50-002-34214 Supplier Medchemexpress LLC Supplier No. HY12022100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminobenzamide is a small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) used in biochemical and cellular research to study DNA repair and oxidant-induced cellular responses. It is supplied as an analytical standard suitable for HPLC, GC, and MS applications.

  • Cas 3544-24-9.
  • Chemical formula C7H8N2O, molecular weight 136.15 g/mol.
  • Used as a PARP inhibitor in biochemical and cellular assays.
  • Supplied as an analytical standard for qualitative and quantitative analysis.
  • Available in multiple pack sizes including 100 MG.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.