Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzamide, 98%

Catalog No. AAA1079306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1079306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1079306 Supplier Thermo Scientific Chemicals Supplier No. A1079306

missing translation for 'validMainframeMsg'

CAS: 3544-24-9 | C7H8N2O | 136.15 g/mol

3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

Solubility
Soluble in dimethylformamide (∼30 mg/ml), dimethyl sulfoxide (∼30 mg/ml), phosphate buffered saline (pH7.2) (∼2 mg/ml), water (25 mg/ml), and ethanol (25 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3544-24-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007989
InChI Key GSCPDZHWVNUUFI-UHFFFAOYSA-N
Synonym m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide
PubChem CID 1645
ChEBI CHEBI:64042
IUPAC Name 3-aminobenzamide
SMILES NC(=O)C1=CC=CC(N)=C1

Specifications

Melting Point 111°C to 115°C
Quantity 5 g
Beilstein 2802373
Solubility Information Soluble in dimethylformamide (∽30mg/ml),dimethyl sulfoxide (∽30mg/ml),phosphate buffered saline (pH7.2) (∽2mg/ml),water (25mg/ml),and ethanol (25mg/ml).
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3-Aminobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-586-9
RTECSNumber CU8992000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.