Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzoic acid hydrochloride, 98%, Thermo Scientific™

Catalog No. AAL0840714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0840714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0840714 Supplier Thermo Scientific Chemicals Supplier No. L0840714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15151-51-6
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012973
InChI Key PTTMMENRLMBXGK-UHFFFAOYSA-N
Synonym 3-aminobenzoic acid hydrochloride, 3-aminobenzoic acid, chloride, 3-aminobenzoic acid hcl, acmc-1bs8f, m-aminobenzoic acid hydrochloride
PubChem CID 3084218
IUPAC Name 3-aminobenzoic acid;hydrochloride
SMILES C1=CC(=CC(=C1)N)C(=O)O.Cl

Specifications

Melting Point ∽286°C (decomposition)
Quantity 25 g
Beilstein 3695285
Sensitivity Light sensitive
Formula Weight 173.6
Percent Purity 98%
Chemical Name or Material 3-Aminobenzoic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-212-5
RTECSNumber DG1225000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.