Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Aminobenzophenone, 98%, Thermo Scientific™
Spécifications
Spécifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 84.0°C to 87.0°C |
| Color | Yellow to Green |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H5C(O)C6H4NH2 |
| Quantity | 1 g |
| Formula Weight | 197.24 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | GHS H Statement: Causes serious eye irritation. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 220-614-4 |
RUO – Research Use Only
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'