Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminobenzothioamide | 78950-36-4 | MFCD04973325 | ≥97.0% | 152.22 | C7H8N2S | 250 MG
SDP

Catalog No. 5000283668
Encompass_Preferred

A small-molecule synthetic intermediate used in pharmaceutical synthesis. It has the chemical formula C7H8N2S and a molecular weight of 152.22 g/mol; it is intended for research and synthesis applications in medicinal chemistry.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • CAS number 78950-36-4.
  • Molecular formula C7H8N2S.
  • Molecular weight 152.22 g/mol.
  • Typical purity ≥97%.
  • Storage: room temperature, protect from light; in solvent: -80°C (2 years), -20°C (1 year).

Catalog No. 50-002-83668 Supplier Medchemexpress LLC Supplier No. HYW128619250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule synthetic intermediate used in pharmaceutical synthesis. It has the chemical formula C7H8N2S and a molecular weight of 152.22 g/mol; it is intended for research and synthesis applications in medicinal chemistry.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • CAS number 78950-36-4.
  • Molecular formula C7H8N2S.
  • Molecular weight 152.22 g/mol.
  • Typical purity ≥97%.
  • Storage: room temperature, protect from light; in solvent: -80°C (2 years), -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.