Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1877-77-6
C7H9NO
123.15
MFCD00007817
OJZQOQNSUZLSMV-UHFFFAOYSA-N
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol
80293
(3-aminophenyl)methanol
C1=CC(=CC(=C1)N)CO
1877-77-6
C7H9NO
123.15
MFCD00007817
OJZQOQNSUZLSMV-UHFFFAOYSA-N
3-aminobenzyl alcohol, 3-aminophenyl methanol, m-aminobenzyl alcohol, 3-aminobenzylalcohol, 3-hydroxymethyl aniline, benzenemethanol, 3-amino, benzyl alcohol, m-amino, 3-aminobenzenemethanol, unii-kn96abq0sp, m-aminophenylcarbinol
80293
(3-aminophenyl)methanol
C1=CC(=CC(=C1)N)CO
Color Beige or Beige-Gray to Brown
Quantity 25g
Melting Point 91.0°C to 95.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula H2NC6H4CH2OH
Beilstein 13, 619
Formula Weight 123.15
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Aminobenzyl alcohol
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 217-514-8