Learn More
Medchemexpress LLC 3-Aminobutanoic acid | 541-48-0 | 98.0% | 103.12 | 25 G
SDP
Catalog No. 5000451367
Encompass_Preferred
3-Aminobutanoic acid is a β-amino acid. It can protect plants against a challenge infection with *P. infestans*, showing various levels of susceptibility for the pathogen. It is for research use only.
  • Increases anthocyanin content in purple basil leaves.
  • Reduces iron ion chelating ability.
  • Enhances the inhibition ability of LOX and COX-2.
  • Activates phenylalanine ammonia-lyase (PAL) and polyphenol oxidase (PPO), and increases their mRNA expression.
  • Reduces the survival rate, pupation rate, and eclosion rate of tea geometrid larvae, and prolongs its larval development period.
  • Induces systemic acquired resistance (SAR) in plants, enhancing plant resistance to pathogens and pests.
Catalog No. 50-004-51367 Supplier Medchemexpress LLC Supplier No. 25GHYW008833
Add to Cart
Add to Cart
3-Aminobutanoic acid is a β-amino acid. It can protect plants against a challenge infection with *P. infestans*, showing various levels of susceptibility for the pathogen. It is for research use only.
  • Increases anthocyanin content in purple basil leaves.
  • Reduces iron ion chelating ability.
  • Enhances the inhibition ability of LOX and COX-2.
  • Activates phenylalanine ammonia-lyase (PAL) and polyphenol oxidase (PPO), and increases their mRNA expression.
  • Reduces the survival rate, pupation rate, and eclosion rate of tea geometrid larvae, and prolongs its larval development period.
  • Induces systemic acquired resistance (SAR) in plants, enhancing plant resistance to pathogens and pests.