Learn More
Medchemexpress LLC 3α-Aminocholestane | 2206-20-4 | 98.0% | 387.68 | 1 ML
SDP
Catalog No. 5000401162
Encompass_Preferred
3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. It is for research use only.
  • Effectively reduces OPM2 cell viability
  • Blocks cell cycle progression in G0/G1 phase in RPMI8226 and U266 cells
  • Reduces multiple myeloma (MM) growth in NOD/SCID/γcIL2R (NSG) mice
  • Significantly enhanced survival of mice after tumor challenge
  • Exhibits antiviral activity against Influenza A/PR8/34 (H1N1) virus in MDCK2 cells
Catalog No. 50-004-01162 Supplier Medchemexpress LLC Supplier No. HY1977610MM/1ML
3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. It is for research use only.
  • Effectively reduces OPM2 cell viability
  • Blocks cell cycle progression in G0/G1 phase in RPMI8226 and U266 cells
  • Reduces multiple myeloma (MM) growth in NOD/SCID/γcIL2R (NSG) mice
  • Significantly enhanced survival of mice after tumor challenge
  • Exhibits antiviral activity against Influenza A/PR8/34 (H1N1) virus in MDCK2 cells