Learn More
Medchemexpress LLC 3α-Aminocholestane | 2206-20-4 | 98.0% | 387.68 | 25 MG
SDP
Catalog No. 5000402461
Encompass_Preferred
3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. It is for research use only and not sold to patients.
  • Appears as a white to off-white solid
  • Stored at -20°C for 3 years or 4°C for 2 years as powder
  • Stored at -80°C for 2 years or -20°C for 1 year in solvent
  • Soluble in ethanol (50 mg/mL), slightly soluble in DMSO
  • Exhibits antiviral activity against Influenza A/PR8/34 (H1N1) virus in MDCK-II cells
  • Reduces OPM2 cell viability
  • Blocks cell cycle progression in G0/G1 phase in RPMI8226 and U266 cells
  • Reduces multiple myeloma (MM) growth in vivo and enhances survival of mice
Catalog No. 50-004-02461 Supplier Medchemexpress LLC Supplier No. 25MGHY19776
3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. It is for research use only and not sold to patients.
  • Appears as a white to off-white solid
  • Stored at -20°C for 3 years or 4°C for 2 years as powder
  • Stored at -80°C for 2 years or -20°C for 1 year in solvent
  • Soluble in ethanol (50 mg/mL), slightly soluble in DMSO
  • Exhibits antiviral activity against Influenza A/PR8/34 (H1N1) virus in MDCK-II cells
  • Reduces OPM2 cell viability
  • Blocks cell cycle progression in G0/G1 phase in RPMI8226 and U266 cells
  • Reduces multiple myeloma (MM) growth in vivo and enhances survival of mice