Learn More
Medchemexpress LLC 3α-Aminocholestane | 2206-20-4 | 98.0% | 387.68 | 5 MG
SDP
Catalog No. 5000402004
Encompass_Preferred
3α-Aminocholestane is a selective inhibitor of SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) with an IC50 of approximately 2.5 μM. It is intended for research use only. This compound effectively reduces multiple myeloma growth in vivo and blocks cell cycle progression.
  • Selectively inhibits SHIP1.
  • Effectively reduces OPM2 cell viability.
  • Blocks cell cycle progression in G0/G1 phase or reduces cells in S phase.
  • Reduces multiple myeloma (MM) growth in vivo.
  • Significantly enhances survival of mice after tumor challenge.
This item is not returnable. View return policy
Catalog No. 50-004-02004 Supplier Medchemexpress LLC Supplier No. HY197765MG
3α-Aminocholestane is a selective inhibitor of SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) with an IC50 of approximately 2.5 μM. It is intended for research use only. This compound effectively reduces multiple myeloma growth in vivo and blocks cell cycle progression.
  • Selectively inhibits SHIP1.
  • Effectively reduces OPM2 cell viability.
  • Blocks cell cycle progression in G0/G1 phase or reduces cells in S phase.
  • Reduces multiple myeloma (MM) growth in vivo.
  • Significantly enhances survival of mice after tumor challenge.