Learn More
Medchemexpress LLC HY-19776 50mg Medchemexpress, 3α-Aminocholestane CAS:2206-20-4 Purity:98%
SDP
Catalog No. 501638936
Encompass_Preferred
Medchemexpress,HY-19776 50mg 3α-Aminocholestane CAS:2206-20-4 Formula:C27H49N 3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8936 Supplier Medchemexpress LLC Supplier No. HY1977650MG
Medchemexpress,HY-19776 50mg 3α-Aminocholestane CAS:2206-20-4 Formula:C27H49N 3α-Aminocholestane is a selective SH2 domain-containing inositol-5′-phosphatase 1 (SHIP1) inhibitor with an IC50 of ~2.5 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.