Learn More
Medchemexpress LLC 3-aminochromen-2-one | 1635-31-0 | 99.2% | 161.16 g·mol⁻¹ | C9H7NO2 | 100 MG
SDP
Catalog No. 5000301293
Encompass_Preferred
3-Aminocoumarin (3-aminochromen-2-one) is a small organic intermediate used in coordination chemistry and materials research. It serves as a ligand precursor for the synthesis of metal complexes and related coordination compounds. Supplied for research use, the compound is offered at high reported purity and in multiple laboratory package sizes.
  • Intermediate for synthesis of metal complexes and coordination compounds.
  • Acts as a ligand precursor in coordination chemistry.
  • High purity suitable for research applications.
  • Available in multiple small-scale package sizes for laboratory use.
  • Molecular formula C9H7NO2; molecular weight 161.16 g·mol⁻¹.
Catalog No. 50-003-01293 Supplier Medchemexpress LLC Supplier No. HYW030266100MG
3-Aminocoumarin (3-aminochromen-2-one) is a small organic intermediate used in coordination chemistry and materials research. It serves as a ligand precursor for the synthesis of metal complexes and related coordination compounds. Supplied for research use, the compound is offered at high reported purity and in multiple laboratory package sizes.
  • Intermediate for synthesis of metal complexes and coordination compounds.
  • Acts as a ligand precursor in coordination chemistry.
  • High purity suitable for research applications.
  • Available in multiple small-scale package sizes for laboratory use.
  • Molecular formula C9H7NO2; molecular weight 161.16 g·mol⁻¹.