Learn More
Medchemexpress LLC 3-aminochromen-2-one | 1635-31-0 | MFCD00016965 | 99.2% | 161.16 | C9H7NO2 | 500 MG
SDP
Catalog No. 5000300248
Encompass_Preferred
3-aminochromen-2-one is a coumarin-derived organic compound used primarily as a synthetic intermediate in coordination chemistry and organic synthesis. It is employed as a building block for the preparation of metal complexes and functionalized coumarin derivatives for research use. Chemical formula C9H7NO2; molecular weight 161.16.
  • Used as an intermediate for synthesis of metal complexes.
  • Can act as a ligand in coordination chemistry.
  • Serves as a building block for fluorescent coumarin derivatives.
  • Suitable for research-scale synthetic applications.
  • Characterized by known molecular formula and molecular weight.
Catalog No. 50-003-00248 Supplier Medchemexpress LLC Supplier No. HYW030266500MG
3-aminochromen-2-one is a coumarin-derived organic compound used primarily as a synthetic intermediate in coordination chemistry and organic synthesis. It is employed as a building block for the preparation of metal complexes and functionalized coumarin derivatives for research use. Chemical formula C9H7NO2; molecular weight 161.16.
  • Used as an intermediate for synthesis of metal complexes.
  • Can act as a ligand in coordination chemistry.
  • Serves as a building block for fluorescent coumarin derivatives.
  • Suitable for research-scale synthetic applications.
  • Characterized by known molecular formula and molecular weight.