Learn More
Medchemexpress LLC 3-aminocoumarin (3-aminochromen-2-one) | 1635-31-0 | MFCD00016965 | 161.16 g/mol | C9H7NO2 | 1 G
SDP
Catalog No. 5000301575
Encompass_Preferred
3-Aminocoumarin (3-aminochromen-2-one) is an organic intermediate commonly used in coordination chemistry and organic synthesis as a precursor to ligands and metal complexes. Supplied for research use, it is intended for preparative and exploratory synthesis workflows where a well-characterized coumarin derivative is required.
  • Serves as an intermediate for synthesis of metal complexes and ligands.
  • Suitable for coordination chemistry and related organic synthesis applications.
  • Characterized small molecule: formula C9H7NO2, molecular weight 161.16 g/mol.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only.
Catalog No. 50-003-01575 Supplier Medchemexpress LLC Supplier No. HYW0302661G
3-Aminocoumarin (3-aminochromen-2-one) is an organic intermediate commonly used in coordination chemistry and organic synthesis as a precursor to ligands and metal complexes. Supplied for research use, it is intended for preparative and exploratory synthesis workflows where a well-characterized coumarin derivative is required.
  • Serves as an intermediate for synthesis of metal complexes and ligands.
  • Suitable for coordination chemistry and related organic synthesis applications.
  • Characterized small molecule: formula C9H7NO2, molecular weight 161.16 g/mol.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only.