Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-aminocoumarin (3-aminochromen-2-one) | 1635-31-0 | MFCD00016965 | 99.2% | 161.16 | C9H7NO2 | 250 MG
SDP

Catalog No. 5000303003
Encompass_Preferred

3-Aminocoumarin (3-aminochromen-2-one) is a coumarin-derived organic intermediate used in research and chemical synthesis. It functions as a building block for the preparation of metal complexes and other functionalized coumarin derivatives, and is supplied as a laboratory research reagent with recommended storage instructions to preserve stability.

  • Coumarin-derived intermediate for organic synthesis.
  • Suitable for synthesis of metal complexes and functionalized derivatives.
  • Molecular formula C9H7NO2; molecular weight 161.16 g/mol.
  • Recommended storage: solid at 4°C, protect from light; solutions -80°C (6 months) or -20°C (1 month).
  • Provided as a research reagent; follow appropriate safety and handling procedures.

Catalog No. 50-003-03003 Supplier Medchemexpress LLC Supplier No. 250MGHYW030266
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminocoumarin (3-aminochromen-2-one) is a coumarin-derived organic intermediate used in research and chemical synthesis. It functions as a building block for the preparation of metal complexes and other functionalized coumarin derivatives, and is supplied as a laboratory research reagent with recommended storage instructions to preserve stability.

  • Coumarin-derived intermediate for organic synthesis.
  • Suitable for synthesis of metal complexes and functionalized derivatives.
  • Molecular formula C9H7NO2; molecular weight 161.16 g/mol.
  • Recommended storage: solid at 4°C, protect from light; solutions -80°C (6 months) or -20°C (1 month).
  • Provided as a research reagent; follow appropriate safety and handling procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.