Learn More
TARGETMOL CHEMICALS INC 3-Aminocoumarin 50MG
SDP
Catalog No. 502880207
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 3-Aminocoumarin (3-aminochromen-2-one) is the key intermediate for the metal complexes. Purity 99.54%
Catalog No. 50-288-0207 Supplier TARGETMOL CHEMICALS INC Supplier No. TN124350MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 3-Aminocoumarin (3-aminochromen-2-one) is the key intermediate for the metal complexes. Purity 99.54%