Learn More
Accela Chembio Inc 3-aminoisonicotinic Acid | 25g | 7579-20-6 | MFCD00137840 | 95+% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon
Catalog No. 50353708
Encompass_Preferred
3-aminoisonicotinic Acid | 25g | 7579-20-6 | MFCD00137840 | 95+% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon
Catalog No. 50-353-708 Supplier Accela Chembio Inc Supplier No. SY00333025G
3-aminoisonicotinic Acid | 25g | 7579-20-6 | MFCD00137840 | 95+% | Shelf Life: 2160 Days | Light Sensitive/nitrogen Or Argon