Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Aminoisonicotinic acid | 7579-20-6 | 97.5% | C6H6N2O2 | 10 G
SDP

Catalog No. 5000434257
Encompass_Preferred

3-Aminoisonicotinic acid is a biochemical reagent that functions as a biological material or organic compound. It is suitable for various life science related research applications and is provided as a solid.

  • Molecular weight of 138.12
  • Purity of 99.73%
  • Light yellow to brown appearance
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-34257 Supplier Medchemexpress LLC Supplier No. HY6525710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Aminoisonicotinic acid is a biochemical reagent that functions as a biological material or organic compound. It is suitable for various life science related research applications and is provided as a solid.

  • Molecular weight of 138.12
  • Purity of 99.73%
  • Light yellow to brown appearance
  • Store at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.