Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. A21331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21331G 1 g
A21335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21331G Supplier TCI America Supplier No. A21331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7579-20-6
Molecular Formula C6H5N2O2
Molecular Weight (g/mol) 137.12
MDL Number MFCD00137840
InChI Key FYEQKMAVRYRMBL-UHFFFAOYSA-M
Synonym 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure
PubChem CID 459531
IUPAC Name 3-aminopyridine-4-carboxylate
SMILES NC1=CN=CC=C1C([O-])=O

Specifications

Melting Point 303°C
Quantity 1 g
Formula Weight 138.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminoisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.