Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Aminomethyl)-Proxyl

Catalog No. 27018025MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-018-025MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 27-018-025MG Supplier Sigma Aldrich Supplier No. 27018025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful spin label for studying biological systems. Used to study molecular diffusion into horse spleen ferritin and has been shown to be an effective nonthiol radioprotector.Used in pharmacological studies of myeloperoxidase-mediated hypochlorous acid production inhibitionReactant involved in: Reduction reactions with nitroxidesSynthesis of adenosine methycarboxamides for use as cardioprotectants

Specifications

CAS 26782-71-8
Melting Point ≥300°C
Molecular Formula C6H13NO2
Linear Formula (CH3)3CCH(NH2)CO2H
MDL Number MFCD00004265
Quantity 500 mg
Synonym 3-(Aminomethyl)-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Molecular Weight (g/mol) 131.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.