Learn More
Medchemexpress LLC 3-(Aminomethyl)pyridine | 3731-52-0 | MFCD00006412 | 108.1411 g/mol | C6H8N2 | 10g
SDP
Catalog No. 5000269577
Encompass_Preferred
3-Picolylamine is a primary amine compound containing a pyridine ring structure 3-Picolylamine inhibits histaminase (diamine oxidase) activity in pig kidneys 3-Picolylamine significantly enhances the contractile response of isolated guinea pig ileum to Histamine (HY-B1204) 3-Picolylamine can be used to study histamine-related physiological mechanisms or allergic diseases (such as histamine-mediated inflammatory responses)[1][2]
Catalog No. 50-002-69577 Supplier Medchemexpress LLC Supplier No. HYY004710G
3-Picolylamine is a primary amine compound containing a pyridine ring structure 3-Picolylamine inhibits histaminase (diamine oxidase) activity in pig kidneys 3-Picolylamine significantly enhances the contractile response of isolated guinea pig ileum to Histamine (HY-B1204) 3-Picolylamine can be used to study histamine-related physiological mechanisms or allergic diseases (such as histamine-mediated inflammatory responses)[1][2]